missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dichlorobenzeneboronic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2429206

 View more versions of this product

Catalog No. AAB2429206



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3,4-Dichlorobenzeneboronic acid
270°C to 274°C
MFCD01074646
7369790
JKIGHOARKAIPJI-UHFFFAOYSA-N
(3,4-dichlorophenyl)boronic acid
2734330
97%
151169-75-4
C6H5BCl2O2
5 g
3,4-dichlorophenyl boronic acid, 3,4-dichlorobenzeneboronic acid, 3,4-dichlorophenylboronicacid, 3,4-dichlorophenyl boranediol, 3,4-dichlorophenylbornic acid, chembl20776, 3,4-dichloro benzene boronic acid, boronic acid, 3,4-dichlorophenyl, pubchem1812
OB(O)C1=CC=C(Cl)C(Cl)=C1
190.81
190.82

Chemical Identifiers

151169-75-4
190.81
JKIGHOARKAIPJI-UHFFFAOYSA-N
2734330
OB(O)C1=CC=C(Cl)C(Cl)=C1
C6H5BCl2O2
MFCD01074646
3,4-dichlorophenyl boronic acid, 3,4-dichlorobenzeneboronic acid, 3,4-dichlorophenylboronicacid, 3,4-dichlorophenyl boranediol, 3,4-dichlorophenylbornic acid, chembl20776, 3,4-dichloro benzene boronic acid, boronic acid, 3,4-dichlorophenyl, pubchem1812
(3,4-dichlorophenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only