Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dichlorobenzamide, 97%

$81.43 - $350.96

Chemical Identifiers

Chemical Name or Material 3,4-Dichlorobenzamide
CAS 2670-38-4
Molecular Formula C7H5Cl2NO
Formula Weight 190.03
MDL Number MFCD00053009
Beilstein 3089065
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1387803

  SDS
alfa aesar™
A1387803
1g Each for $81.43

AAA1387806

  SDS
alfa aesar™
A1387806
5g Each for $350.96
Description & Specifications

Specifications

Formula Weight 190.03
Chemical Name or Material 3,4-Dichlorobenzamide
Assay 97%
Molecular Formula C7H5Cl2NO
MDL Number MFCD00053009
CAS 2670-38-4
Beilstein 3089065
Melting Point 140° to 142°C
Quantity 1g