Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dichlorobenzaldoxime, 98%

$62.34 - $217.04

Chemical Identifiers

Chemical Name or Material 3,4-Dichlorobenzaldoxime
CAS 5331-92-0
Molecular Formula C7H5Cl2NO
Formula Weight 190.03
MDL Number MFCD00017592
Beilstein 2206792
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1020203

 
alfa aesar™
L1020203
1g Each for $62.34

AAL1020206

 
alfa aesar™
L1020206
5g Each for $217.04
Description & Specifications

Specifications

Formula Weight 190.03
Chemical Name or Material 3,4-Dichlorobenzaldoxime
Molecular Formula C7H5Cl2NO
MDL Number MFCD00017592
Beilstein 2206792
Assay 98%
CAS 5331-92-0
Melting Point 121° to 123°C
Quantity 1g