missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dichloroanisole, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 264191000

 View more versions of this product

Catalog No. AC264191000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥99.0%
Liquid
Clear light yellow
99.0
Authentic
36404-30-5
C7H6Cl2O
99+%
Cl2C6H3OCH3
177.03
100g
100.0
1.5560 to 1.5580 (20°C, 589nm)
1, 2-Dichloro-4-methoxybenzene
99+%
MFCD00070785
3, 4-Dichloroanisole

Chemical Identifiers

36404-30-5
100.0
C7H6Cl2O
177.03
1, 2-Dichloro-4-methoxybenzene
≥99.0%
99.0
99+%
Cl2C6H3OCH3
MFCD00070785
3, 4-Dichloroanisole, 99+%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA