Learn More
3,4-Dichloroanisole, 99+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 264190250
Specifications
3, 4-Dichloroanisole | |
Glass bottle | |
Yellow | |
99+% | |
99.0 | |
99% min. (GC) | |
Cl2C6H3OCH3 | |
1,2-dichloro-4-methyxybenzene, 3,4-dichloroanisole, 3,4-dichloroanizole, 3,4-dichloromethoxybenzene, 3,4-dichlorophenol, methyl ether, 4,5-dichlorophenyl methyl ether, acmc-1ajco, benzene, 1,2-dichloro-4-methoxy, pubchem3683 | |
COC1=CC(=C(C=C1)Cl)Cl | |
177.03 | |
177.03 | |
99+% |
Authentic | |
1.5560 to 1.558 | |
25g | |
36404-30-5 | |
100.0 | |
C7H6Cl2O | |
MFCD00070785 | |
VISJRVXHPNMYRH-UHFFFAOYSA-N | |
1,2-dichloro-4-methoxybenzene | |
37438 | |
Liquid |
Chemical Identifiers
36404-30-5 | |
177.03 | |
VISJRVXHPNMYRH-UHFFFAOYSA-N | |
37438 | |
COC1=CC(=C(C=C1)Cl)Cl |
C7H6Cl2O | |
MFCD00070785 | |
1,2-dichloro-4-methyxybenzene, 3,4-dichloroanisole, 3,4-dichloroanizole, 3,4-dichloromethoxybenzene, 3,4-dichlorophenol, methyl ether, 4,5-dichlorophenyl methyl ether, acmc-1ajco, benzene, 1,2-dichloro-4-methoxy, pubchem3683 | |
1,2-dichloro-4-methoxybenzene |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
TSCA : TSCA