Please login to your online account to display your discounted pricing

3,4-Dichloroanisole, 99+%, ACROS Organics™

$256.19 - $256.19

Chemical Identifiers

Chemical Name or Material 3, 4-Dichloroanisole
Name Note 99+%
CAS 36404-30-5
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C7H6Cl2O
Linear Formula Cl2C6H3OCH3
Formula Weight 177.03
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC264190250

  SDS
thermo fisher scientific
264190250
Glass bottle 25g Each for $256.19

AC264191000

  SDS
thermo fisher scientific
264191000
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.0%
Physical Form Liquid
Formula Weight 177.03
Quantity 25g
Color Clear light yellow
TSCA TSCA
CAS Min % 99.0
CAS Max % 100.0
Refractive Index 1.5560 to 1.5580 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Chemical Name or Material 3, 4-Dichloroanisole
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 99+%
Assay Percent Range 99+%
Molecular Formula C7H6Cl2O
Linear Formula Cl2C6H3OCH3
MDL Number MFCD00070785
CAS 36404-30-5
Synonym 1, 2-Dichloro-4-methoxybenzene
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.