Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dichloroanisole, 98%

$934.19 - $934.19

Chemical Identifiers

Chemical Name or Material 3,4-Dichloroanisole
CAS 36404-30-5
Molecular Formula C7H6Cl2O
Formula Weight 177.03
MDL Number MFCD00070785
Beilstein 775080
Synonym 3,4-Dichloromethoxybenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1724222

  SDS
alfa aesar™
A1724222
100g Each for $934.19
Description & Specifications

Specifications

Formula Weight 177.03
Chemical Name or Material 3,4-Dichloroanisole
Assay 98%
Molecular Formula C7H6Cl2O
MDL Number MFCD00070785
CAS 36404-30-5
Beilstein 775080
Synonym 3,4-Dichloromethoxybenzene
Flash Point >110°C (230°F)
Boiling Point 200° to 204°C
Melting Point −8°C
Quantity 100g