Learn More
Thermo Scientific Chemicals 3,4-Dichloroaniline, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 113110050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3, 4-Dichloroaniline | |
95-76-1 | |
100.0 | |
69.0°C to 73.0°C | |
166°C | |
Authentic | |
Cl2C6H3NH2 | |
12, 626 | |
15, 3063 | |
3,4-dichlorobenzenamine, 3,4-dichloraniline, 3,4-dichloranilin, 4,5-dichloroaniline, benzenamine, 3,4-dichloro, 1-amino-3,4-dichlorobenzene, 3,4-dca, 3,4-dichlorophenylamine, aniline, 3,4-dichloro, m,p-dichloroaniline | |
SDYWXFYBZPNOFX-UHFFFAOYSA-N | |
3,4-dichloroaniline | |
7257 | |
162.02 | |
98% |
98% | |
97.5 | |
Beige to Brown | |
272.0°C | |
97.5% min. (GC) | |
C6H5Cl2N | |
MFCD00007768 | |
Glass bottle | |
5g | |
Solubility in water: 0.06g/100mL in water. Other solubilities: soluble in most common organic solvents,very soluble in alcohol and ether,slightly soluble in benzene | |
C1=CC(=C(C=C1N)Cl)Cl | |
162.02 | |
CHEBI:16767 | |
Crystalline Mass and/or Chunks |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
May cause an allergic skin reaction.
Toxic in contact with skin.
Causes serious eye damage.
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Gently wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 202-448-4
RTECSNumber : BX2625000
TSCA : TSCA
Recommended Storage : Product darkens on storage
RUO â Research Use Only