missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dichloroaniline, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 113110010

Catalog No. AC113110010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3, 4-Dichloroaniline
95-76-1
100.0
166°C
97.5% min. (GC)
C6H5Cl2N
MFCD00007768
12, 626
3,4-dichlorobenzenamine, 3,4-dichloraniline, 3,4-dichloranilin, 4,5-dichloroaniline, benzenamine, 3,4-dichloro, 1-amino-3,4-dichlorobenzene, 3,4-dca, 3,4-dichlorophenylamine, aniline, 3,4-dichloro, m,p-dichloroaniline
SDYWXFYBZPNOFX-UHFFFAOYSA-N
3,4-dichloroaniline
7257
162.02
98%
97.5
272.0°C
Authentic
Glass bottle
Cl2C6H3NH2
1kg
15, 3063
Solubility in water: 0.06g/100mL in water. Other solubilities: soluble in most common organic solvents,very soluble in alcohol and ether,slightly soluble in benzene
C1=CC(=C(C=C1N)Cl)Cl
162.02
CHEBI:16767
98%
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
May cause an allergic skin reaction.
Toxic in contact with skin.
Causes serious eye damage.
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Gently wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 202-448-4

RTECSNumber : BX2625000

TSCA : TSCA

Recommended Storage : Product darkens on storage

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only