Learn More
Alfa Aesar™ 3,4-Dichloroaniline, 98%
Manufacturer: Alfa Aesar™ A1519936
Specifications
3,4-Dichloroaniline | |
272°C | |
500g | |
C6H5Cl2N | |
UN3442 | |
14,3054 | |
SDYWXFYBZPNOFX-UHFFFAOYSA-N | |
3,4-dichloroaniline | |
7257 | |
162.02 |
135°C (275°F) | |
70°C to 73°C | |
95-76-1 | |
MFCD00007768 | |
636837 | |
3,4-dichlorobenzenamine, 3,4-dichloraniline, 3,4-dichloranilin, 4,5-dichloroaniline, benzenamine, 3,4-dichloro, 1-amino-3,4-dichlorobenzene, 3,4-dca, 3,4-dichlorophenylamine, aniline, 3,4-dichloro, m,p-dichloroaniline | |
C1=CC(=C(C=C1N)Cl)Cl | |
162.013 | |
CHEBI:16767 | |
98% |
Chemical Identifiers
95-76-1 | |
162.013 | |
SDYWXFYBZPNOFX-UHFFFAOYSA-N | |
7257 | |
3,4-dichloroaniline |
C6H5Cl2N | |
MFCD00007768 | |
3,4-dichlorobenzenamine, 3,4-dichloraniline, 3,4-dichloranilin, 4,5-dichloroaniline, benzenamine, 3,4-dichloro, 1-amino-3,4-dichlorobenzene, 3,4-dca, 3,4-dichlorophenylamine, aniline, 3,4-dichloro, m,p-dichloroaniline | |
CHEBI:16767 | |
C1=CC(=C(C=C1N)Cl)Cl |
Safety and Handling
GHS H Statement
H310-H330-H318-H302-H317
Fatal in contact with skin.
Fatal if inhaled.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
GHS P Statement
P260-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DICHLOROANILINES, SOLID
RTECSNumber : BX2625000
TSCA : TSCA