Please login to your online account to display your discounted pricing

3',4'-Dichloroacetophenone, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3';, 4';-Dichloroacetophenone
Name Note 99%
CAS 2642-63-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8 H6 Cl2 O
Linear Formula Cl2C6H3COCH3
Formula Weight 189.04
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC170951000

  SDS
thermo fisher scientific
170951000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder or Crystals
Assay ≥98.5%
Formula Weight 189.04
Color White to cream
Quantity 100g
EINECS Number 220-146-0
TSCA TSCA
Melting Point 72° to 76°C
Beilstein 07, II, 219
CAS Max % 100.0
Chemical Name or Material 3';, 4';-Dichloroacetophenone
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 99%
Assay Percent Range 99%
Molecular Formula C8 H6 Cl2 O
Linear Formula Cl2C6H3COCH3
MDL Number MFCD00000553
CAS 2642-63-9
Synonym 1-(3, 4-Dichlorophenyl)ethanone
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.