Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dichloro-N-methylaniline, 95%

Chemical Identifiers

Chemical Name or Material 3,4-Dichloro-N-methylaniline
CAS 40750-59-2
Molecular Formula C7H7Cl2N
Formula Weight 176.05g/mol
MDL Number MFCD00052850
UN Number UN2810
Beilstein 2962410
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1063103

 
alfa aesar™
L10631-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1063106

 
alfa aesar™
L10631-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1g
Boiling Point 150° to 152°C (15mmHg)
Quantity 1g
Chemical Name or Material 3,4-Dichloro-N-methylaniline
CAS 40750-59-2
Assay 95%
Molecular Formula C7H7Cl2N
Formula Weight 176.05g/mol
MDL Number MFCD00052850
UN Number UN2810
Beilstein 2962410
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.