Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dichloro-L-phenylalanine, 98%

Chemical Identifiers

Chemical Name or Material 3,4-Dichloro-L-phenylalanine
CAS 52794-99-7
Formula Weight 234.08
MDL Number MFCD00191203
Synonym H-Phe(3,4-Cl2)-OH; (S)-2-Amino-3-(3,4-dichlorophenyl)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6383603

  SDS
alfa aesar™
H6383603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6383606

  SDS
alfa aesar™
H6383606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 234.08
Assay 98%
CAS 52794-99-7
Synonym H-Phe(3,4-Cl2)-OH; (S)-2-Amino-3-(3,4-dichlorophenyl)propionic acid
Chemical Name or Material 3,4-Dichloro-L-phenylalanine
Quantity 1g
Storage Note 1 2° to 8°C
MDL Number MFCD00191203
Physical Form Solid
Safety and Handling

2° to 8°C