missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dichloro-L-phenylalanine, 98%

Manufacturer:  Alfa Aesar™ H6383606

 View more versions of this product

Catalog No. AAH6383606


 

Specifications

Specifications

3,4-Dichloro-L-phenylalanine
98%
MFCD00191203
NRCSJHVDTAAISV-QMMMGPOBSA-N
(2S)-2-amino-3-(3,4-dichlorophenyl)propanoic acid
2761498
Solid
52794-99-7
C9H9Cl2NO2
H-Phe(3,4-Cl2)-OH; (S)-2-Amino-3-(3,4-dichlorophenyl)propionic acid
C1=CC(=C(C=C1CC(C(=O)O)N)Cl)Cl
234.076
234.08
5g

Chemical Identifiers

52794-99-7
234.076
NRCSJHVDTAAISV-QMMMGPOBSA-N
2761498
C9H9Cl2NO2
MFCD00191203
H-Phe(3,4-Cl2)-OH; (S)-2-Amino-3-(3,4-dichlorophenyl)propionic acid
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents