Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dichloro-beta-nitrostyrene, 98+%

$116.80 - $116.80

Chemical Identifiers

Chemical Name or Material 3,4-Dichloro-beta-nitrostyrene
CAS 18984-16-2
Molecular Formula C8H5Cl2NO2
Formula Weight 218.04g/mol
MDL Number MFCD00052834
Beilstein 2211292
Synonym 1-(3,4-Dichlorophenyl)-2-nitroethene; 1-(3,4-Dichlorophenyl)-2-nitroethylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1517006

 
alfa aesar™
A15170-06
5g Each for $116.80
Description & Specifications

Specifications

Melting Point 90° to 95°C
Quantity 5g
Chemical Name or Material 3,4-Dichloro-beta-nitrostyrene
CAS 18984-16-2
Assay ≥98%
Molecular Formula C8H5Cl2NO2
Formula Weight 218.04g/mol
MDL Number MFCD00052834
Beilstein 2211292
Synonym 1-(3,4-Dichlorophenyl)-2-nitroethene; 1-(3,4-Dichlorophenyl)-2-nitroethylene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.