missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dichloro-1,2,5-thiadiazole, 99%

Manufacturer:  Alfa Aesar™ A1173809

 View more versions of this product

Catalog No. AAA1173809



Description

Description

Applications
3,4-Dichloro-1,2,5-thiadiazole is a chlorinated thiadiazole that is a potent inhibitors of nitrification in soil. 3,4-Dichloro-1,2,5-thiadiazole is used as a building block in the preparaton of the β-adrenergic blocking agent, Timolol.

Solubility
Soluble in chloroform and methanol.

Notes
Hygroscopic. Store under dry inert gas. Incompatible with moisture and oxidizing agents, acids and bases.
Specifications

Specifications

3,4-Dichloro-1,2,5-thiadiazole
157°C to 159°C
5728-20-1
MFCD00010072
606585
YNZQOVYRCBAMEU-UHFFFAOYSA-N
3,4-dichloro-1,2,5-thiadiazole
79804
99%
>100°C (212°F)
10g
C2Cl2N2S
UN2810
1,2,5-thiadiazole, 3,4-dichloro, dichloro-1,2,5-thiadiazole, 3,4-dichloro-1,2,5 thiadiazole, pubchem20790, acmc-1al9l, ksc272e3d, 4,5-dichloro-2,1,3-thiadiazole, 3,4-bis chloranyl-1,2,5-thiadiazole, 3,4-dichloro-1,2,5-thiadiazole
C1(=NSN=C1Cl)Cl
154.996
155.01

Chemical Identifiers

5728-20-1
154.996
YNZQOVYRCBAMEU-UHFFFAOYSA-N
79804
C1(=NSN=C1Cl)Cl
C2Cl2N2S
MFCD00010072
1,2,5-thiadiazole, 3,4-dichloro, dichloro-1,2,5-thiadiazole, 3,4-dichloro-1,2,5 thiadiazole, pubchem20790, acmc-1al9l, ksc272e3d, 4,5-dichloro-2,1,3-thiadiazole, 3,4-bis chloranyl-1,2,5-thiadiazole, 3,4-dichloro-1,2,5-thiadiazole
3,4-dichloro-1,2,5-thiadiazole
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

SDS Documents