missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dibromothiophene, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC264520050

 View more versions of this product

Catalog No. AC264520050



Specifications

Specifications

3, 4-Dibromothiophene
Authentic
1.6390 to 1.6410 (20°C, 589nm)
5g
96.0
97%
MFCD00005465
VGKLVWTVCUDISO-UHFFFAOYSA-N
3,4-dibromothiophene
18452
Liquid
2.1300g/mL
Glass bottle
Colorless to Yellow
3141-26-2
100.0
C4H2Br2S
thiophene, 3,4-dibromo, 3,4-dibromo-thiophene, pubchem5510, thiophene,4-dibromo, 3, 4-dibromothiophene, acmc-1clwy, 3,4-bis bromanyl thiophene, ksc224a6j, 3,4-dibromothiophene, bidd:gt0493
C1=C(C(=CS1)Br)Br
241.928
241.94
≥96.0%

Chemical Identifiers

3141-26-2
241.928
VGKLVWTVCUDISO-UHFFFAOYSA-N
18452
C1=C(C(=CS1)Br)Br
C4H2Br2S
MFCD00005465
thiophene, 3,4-dibromo, 3,4-dibromo-thiophene, pubchem5510, thiophene,4-dibromo, 3, 4-dibromothiophene, acmc-1clwy, 3,4-bis bromanyl thiophene, ksc224a6j, 3,4-dibromothiophene, bidd:gt0493
3,4-dibromothiophene
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Recommended Storage : May darken on storage

SDS Documents