missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,4-Dibromothiophene, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 264520010
Specifications
3, 4-Dibromothiophene | |
>110°C | |
Glass bottle | |
2.13 | |
Colorless to Yellow | |
1g | |
96.0 | |
96% min. (GC) | |
MFCD00005465 | |
VGKLVWTVCUDISO-UHFFFAOYSA-N | |
3,4-dibromothiophene | |
18452 | |
Liquid |
2.1300g/mL | |
Authentic | |
1.6390 to 1.641 | |
221°C to 222°C | |
4°C to 5°C | |
3141-26-2 | |
100.0 | |
C4H2Br2S | |
3, 4-dibromothiophene, 3,4-bis bromanyl thiophene, 3,4-dibromo-thiophene, 3,4-dibromothiophene, acmc-1clwy, bidd:gt0493, ksc224a6j, pubchem5510, thiophene, 3,4-dibromo, thiophene,4-dibromo | |
C1=C(C(=CS1)Br)Br | |
241.94 | |
241.94 | |
97% |
Chemical Identifiers
3141-26-2 | |
241.94 | |
VGKLVWTVCUDISO-UHFFFAOYSA-N | |
18452 | |
C1=C(C(=CS1)Br)Br |
C4H2Br2S | |
MFCD00005465 | |
3, 4-dibromothiophene, 3,4-bis bromanyl thiophene, 3,4-dibromo-thiophene, 3,4-dibromothiophene, acmc-1clwy, bidd:gt0493, ksc224a6j, pubchem5510, thiophene, 3,4-dibromo, thiophene,4-dibromo | |
3,4-dibromothiophene |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Warning
EINECSNumber : 221-546-8
Recommended Storage : May darken on storage