missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,4-Dibenzyloxybenzaldehyde, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC112640250
Specifications
3, 4-Dibenzyloxybenzaldehyde | |
Glass bottle | |
88°C to 92°C | |
5447-02-9 | |
100.0 | |
C21H18O3 | |
MFCD00004776 | |
XDDLXZHBWVFPRG-UHFFFAOYSA-N | |
79526 | |
Powder |
Authentic | |
White to Yellow | |
25g | |
98.5 | |
98.5% min. (GC) | |
(C6H5CH2O)2C6H3CHO | |
3,4-bis benzyloxy benzaldehyde, 3,4-bis phenylmethoxy benzaldehyde, 3,4-dibenzyl-oxybenzaldehyde, 3,4-dibenzyloxy benzaldehyde, 3,4-dibenzyloxy-benzaldehyde, 3,4-dibenzyloxybenzaldehyde, 3.4-dibenzyloxybenzaldehyde, benzaldehyde, 3,4-dibenzyloxy, xddlxzhbwvfprg-uhfffaoysa, xddlxzhbwvfprg-uhfffaoysa-n | |
318.36 | |
318.36 | |
99% |
Chemical Identifiers
5447-02-9 | |
318.36 | |
XDDLXZHBWVFPRG-UHFFFAOYSA-N | |
79526 |
C21H18O3 | |
MFCD00004776 | |
3,4-bis benzyloxy benzaldehyde, 3,4-bis phenylmethoxy benzaldehyde, 3,4-dibenzyl-oxybenzaldehyde, 3,4-dibenzyloxy benzaldehyde, 3,4-dibenzyloxy-benzaldehyde, 3,4-dibenzyloxybenzaldehyde, 3.4-dibenzyloxybenzaldehyde, benzaldehyde, 3,4-dibenzyloxy, xddlxzhbwvfprg-uhfffaoysa, xddlxzhbwvfprg-uhfffaoysa-n |
Safety and Handling
EINECSNumber : 226-662-2