missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dibenzyloxybenzaldehyde, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC112640050

 View more versions of this product

Catalog No. AC112640050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3, 4-Dibenzyloxybenzaldehyde
Glass bottle
88°C to 92°C
5447-02-9
100.0
C21H18O3
MFCD00004776
XDDLXZHBWVFPRG-UHFFFAOYSA-N
79526
Powder
Authentic
White to Yellow
5g
98.5
98.5% min. (GC)
(C6H5CH2O)2C6H3CHO
3,4-bis benzyloxy benzaldehyde, 3,4-bis phenylmethoxy benzaldehyde, 3,4-dibenzyl-oxybenzaldehyde, 3,4-dibenzyloxy benzaldehyde, 3,4-dibenzyloxy-benzaldehyde, 3,4-dibenzyloxybenzaldehyde, 3.4-dibenzyloxybenzaldehyde, benzaldehyde, 3,4-dibenzyloxy, xddlxzhbwvfprg-uhfffaoysa, xddlxzhbwvfprg-uhfffaoysa-n
318.36
318.36
99%

Chemical Identifiers

5447-02-9
318.36
XDDLXZHBWVFPRG-UHFFFAOYSA-N
79526
C21H18O3
MFCD00004776
3,4-bis benzyloxy benzaldehyde, 3,4-bis phenylmethoxy benzaldehyde, 3,4-dibenzyl-oxybenzaldehyde, 3,4-dibenzyloxy benzaldehyde, 3,4-dibenzyloxy-benzaldehyde, 3,4-dibenzyloxybenzaldehyde, 3.4-dibenzyloxybenzaldehyde, benzaldehyde, 3,4-dibenzyloxy, xddlxzhbwvfprg-uhfffaoysa, xddlxzhbwvfprg-uhfffaoysa-n
Safety and Handling

Safety and Handling

EINECSNumber : 226-662-2

SDS Documents