missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[3,4]-Dibenzo-21-crown-7, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC278160010

 View more versions of this product

Catalog No. AC278160010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Chelation/Complexation reagent
Specifications

Specifications

[3,4]-Dibenzo-21-crown-7
Glass bottle
104.0°C to 108.0°C
98%
97.5
98%
MFCD00068646
JKCQOMAQPUYHPL-UHFFFAOYSA-N
404.45
404.45
98%
Authentic
White
1g
14098-41-0
100.0
C22H28O7
2,3,14,15-dibenzo-1,4,7,10,13,16,19-heptaoxacycloheneicosa-2,14-diene, 2,5,8,15,18,21,24-heptaoxatricyclo 23.4.0.0?, 1 ? nonacosa-1 29 ,9,11,13,25,27-hexaene, 6,7,9,10,12,13,20,21,23,24-decahydrodibenzo b,k 1,4,7,10,13,16,19 heptaoxacyclohenicosine, 6,7,9,10,12,13,20,21,23,24-decahydrodibenzo b,k 1,4,7,10,13,16,19 heptaoxacyclohenicosine #, dibenzo b,k 1,4,7,10,13,16,19 heptaoxacycloheneicosin,6,7,9,10,12,13,20,21,23,24-decahydro, dibenzo-21-crown, dibenzo-21-crown 7-ether, dibenzo-21-crown-7, dibenzo-21-crown7-ether
C1COCCOC2=CC=CC=C2OCCOCCOC3=CC=CC=C3OCCO1
585932
Fluffy Amorphous Powder

Chemical Identifiers

14098-41-0
404.45
JKCQOMAQPUYHPL-UHFFFAOYSA-N
585932
C22H28O7
MFCD00068646
2,3,14,15-dibenzo-1,4,7,10,13,16,19-heptaoxacycloheneicosa-2,14-diene, 2,5,8,15,18,21,24-heptaoxatricyclo 23.4.0.0?, 1 ? nonacosa-1 29 ,9,11,13,25,27-hexaene, 6,7,9,10,12,13,20,21,23,24-decahydrodibenzo b,k 1,4,7,10,13,16,19 heptaoxacyclohenicosine, 6,7,9,10,12,13,20,21,23,24-decahydrodibenzo b,k 1,4,7,10,13,16,19 heptaoxacyclohenicosine #, dibenzo b,k 1,4,7,10,13,16,19 heptaoxacycloheneicosin,6,7,9,10,12,13,20,21,23,24-decahydro, dibenzo-21-crown, dibenzo-21-crown 7-ether, dibenzo-21-crown-7, dibenzo-21-crown7-ether
C1COCCOC2=CC=CC=C2OCCOCCOC3=CC=CC=C3OCCO1
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA

SDS Documents