Please login to your online account to display your discounted pricing

[3,4]-Dibenzo-21-crown-7, 98%, ACROS Organics™

$373.97 - $373.97

Chemical Identifiers

Chemical Name or Material [3, 4]-Dibenzo-21-crown-7
Name Note 98%
CAS 14098-41-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C22H28O7
Formula Weight 404.45
MDL Number MFCD00068646
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC278160050

  SDS
thermo fisher scientific
AC278160050
Glass bottle 5g Each for $373.97

AC278160010

  SDS
thermo fisher scientific
AC278160010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC278165000

  SDS
thermo fisher scientific
278165000
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Fluffy Amorphous Powder
Assay ≥97.5%
Formula Weight 404.45
Quantity 5g
Color White
TSCA TSCA
Melting Point 104° to 108°C
CAS Max % 100.0
Chemical Name or Material [3, 4]-Dibenzo-21-crown-7
Name Note 98%
Assay Percent Range 98%
Molecular Formula C22H28O7
MDL Number MFCD00068646
CAS 14098-41-0
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle

Chelation/Complexation reagent

Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.