Please login to your online account to display your discounted pricing

[3,4]-Dibenzo-21-crown-7, 98%, ACROS Organics™


Quantity 500mg

Manufacturer: thermo fisher scientific  278165000

 View more versions of this product

Catalog No. AC278165000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Fluffy Amorphous Powder
Formula Weight 404.45
Quantity 500mg
Color White
TSCA TSCA
Melting Point 104° to 108°C
CAS Max % 100.0
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material [3, 4]-Dibenzo-21-crown-7
Name Note 98%
Assay Percent Range 98%
Molecular Formula C22H28O7
MDL Number MFCD00068646
CAS 14098-41-0
Infrared Spectrum Authentic

Specifications

Chemical Name or Material [3, 4]-Dibenzo-21-crown-7
Name Note 98%
CAS 14098-41-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C22H28O7
Formula Weight 404.45
MDL Number MFCD00068646
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents