missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Diaminotoluene, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 112431000

 View more versions of this product

Catalog No. AC112431000



Description

Description

Specifications

Specifications

3, 4-Diaminotoluene
Glass bottle
Brown to Brown-Gray
100g
96.0
96% min. (HPLC)
CH3C6H3(NH2)2
13, 148
Solubility in water: 16g/L (20°C).
CC1=CC(=C(C=C1)N)N
122.17
122.17
97%
Authentic
265.0°C
83.0°C to 90.0°C
496-72-0
100.0
C7H10N2
MFCD00007728
1,2-benzenediamine, 4-methyl, 1,2-diamino-4-methylbenzene, 3,4-diaminotoluene, 3,4-toluenediamine, 3,4-toluylenediamine, 3,4-tolylenediamine, 4-methyl-1,2-benzenediamine, 4-methyl-1,2-phenylenediamine, 4-methyl-o-phenylenediamine, toluene-3,4-diamine
DGRGLKZMKWPMOH-UHFFFAOYSA-N
4-methylbenzene-1,2-diamine
10332
Crystalline Chunks or Flakes

Chemical Identifiers

496-72-0
122.17
DGRGLKZMKWPMOH-UHFFFAOYSA-N
10332
CC1=CC(=C(C=C1)N)N
C7H10N2
MFCD00007728
1,2-benzenediamine, 4-methyl, 1,2-diamino-4-methylbenzene, 3,4-diaminotoluene, 3,4-toluenediamine, 3,4-toluylenediamine, 3,4-tolylenediamine, 4-methyl-1,2-benzenediamine, 4-methyl-1,2-phenylenediamine, 4-methyl-o-phenylenediamine, toluene-3,4-diamine
4-methylbenzene-1,2-diamine
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Suspected of causing genetic defects.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if y

GHS Signal Word: Warning

EINECSNumber : 207-826-2

SDS Documents