Please login to your online account to display your discounted pricing

3,4-Diaminopyridine, 98%, ACROS Organics™

$180.19 - $180.19

Chemical Identifiers

Chemical Name or Material 3, 4-Diaminopyridine
CAS 54-96-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H7N3
Linear Formula (C5H3N)(NH2)2
Formula Weight 109.13
MDL Number MFCD00006401
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC112390250

  SDS
thermo fisher scientific
AC112390250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC112390050

  SDS
thermo fisher scientific
112390050
Glass bottle 5g Each for $180.19
Description & Specifications

Specifications

Melting Point 216° to 221°C (dec.)
Formula Weight 109.13
Physical Form Crystalline Powder
Color Brownish to brown-gray
Assay ≥97.5%
Quantity 25g
Beilstein 22, II, 395
Merck Index 15, 398
CAS Max % 100.0
Linear Formula (C5H3N)(NH2)2
MDL Number MFCD00006401
Assay Percent Range 98%
Molecular Formula C5H7N3
CAS 54-96-6
Health Hazard 3 GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray.
Packaging Glass bottle
CAS Min % 97.5
Chemical Name or Material 3, 4-Diaminopyridine
Infrared Spectrum Authentic
Storage Information Product darkens on storage
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS P Statement Fatal if swallowed. Toxic in contact with skin. Fatal if inhaled. Causes skin irritation. Causes serious eye damage. May cause respiratory irritation.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray.

GHS P Statement Fatal if swallowed. Toxic in contact with skin. Fatal if inhaled. Causes skin irritation. Causes serious eye damage. May cause respiratory irritation.