Learn More
3,4-Diaminopyridine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 112390050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3, 4-Diaminopyridine | |
97.5 | |
216°C to 221°C | |
240°C | |
97.5% min. (GC) | |
C5H7N3 | |
MFCD00006401 | |
22, II, 395 | |
3,4-diaminopyridine, amifampridine, 3,4-pyridinediamine, firdapse, diamino-3,4 pyridine, 4,5-diaminopyridine, 3,4-dap, amifampridin, zenas, pyridine, 3,4-diamino | |
OYTKINVCDFNREN-UHFFFAOYSA-N | |
pyridine-3,4-diamine | |
5918 | |
98% |
54-96-6 | |
100.0 | |
Brown to Brown-Gray | |
Authentic | |
Glass bottle | |
(C5H3N)(NH2)2 | |
5 g | |
15, 398 | |
Solubility in water: 24g/L (20°C) | |
C1=CN=CC(=C1N)N | |
109.13 | |
109.13 | |
Crystalline Powder |
Chemical Identifiers
54-96-6 | |
109.13 | |
OYTKINVCDFNREN-UHFFFAOYSA-N | |
5918 | |
C1=CN=CC(=C1N)N |
C5H7N3 | |
MFCD00006401 | |
3,4-diaminopyridine, amifampridine, 3,4-pyridinediamine, firdapse, diamino-3,4 pyridine, 4,5-diaminopyridine, 3,4-dap, amifampridin, zenas, pyridine, 3,4-diamino | |
pyridine-3,4-diamine |
Safety and Handling
GHS H Statement
Fatal if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection
GHS Signal Word: Danger
EINECSNumber : 200-220-9
Recommended Storage : Product darkens on storage
RUO – Research Use Only