missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Diaminobenzophenone, 99%, ACROS Organics™

Chemical Identifiers

CAS 39070-63-8
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C13H12N2O
Linear Formula C6H5COC6H3(NH2)2
Formula Weight 212.25
MDL Number MFCD00007727
Synonym (3, 4-Diaminophenyl)phenylmethanone
Chemical Name or Material 3, 4-Diaminobenzophenone, 99%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC184800250 SDS Fisher Scientific Acros
184800250
Plastic bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC184801000 SDS Thermo Fisher Scientific
184801000
Plastic bottle 100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

39070-63-8
100.0
C13H12N2O
212.25
(3, 4-Diaminophenyl)phenylmethanone
≥98.5%
98.5
99%
C6H5COC6H3(NH2)2
MFCD00007727
3, 4-Diaminobenzophenone, 99%

Specifications

Yellow to ochre-yellow
25g
98.5
Authentic
Plastic bottle
Powder
C13H12N2O
212.25
14, II, 225
3, 4-Diaminobenzophenone, 99%
116° to 119°C
39070-63-8
100.0
0.5% max.
0.3% max.
99%
C6H5COC6H3(NH2)2
MFCD00007727
(3, 4-Diaminophenyl)phenylmethanone
≥98.5%

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 254-273-8

TSCA : TSCA