Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Diaminobenzophenone, 97%

$28.77 - $232.04

Chemical Identifiers

Chemical Name or Material 3,4-Diaminobenzophenone
CAS 39070-63-8
Molecular Formula C13H12N2O
Formula Weight 212.25
MDL Number MFCD00007727
Beilstein 2212487
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1386306

  SDS
alfa aesar™
A1386306
5g Each for $28.77

AAA1386314

  SDS
alfa aesar™
A1386314
25g Each for $76.54

AAA1386322

  SDS
alfa aesar™
A1386322
100g Each for $232.04
Description & Specifications

Specifications

Formula Weight 212.25
Molecular Formula C13H12N2O
MDL Number MFCD00007727
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 115° to 119°C
Chemical Name or Material 3,4-Diaminobenzophenone
Assay 97%
CAS 39070-63-8
Beilstein 2212487
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.