Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Diaminobenzhydrazide, 98%

$89.80 - $331.49

Chemical Identifiers

Chemical Name or Material 3,4-Diaminobenzhydrazide
CAS 103956-09-8
Molecular Formula C7H10N4O
Formula Weight 166.18
MDL Number MFCD00017069
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0526303

  SDS
alfa aesar™
L0526303
1g Each for $89.80

AAL0526306

  SDS
alfa aesar™
L0526306
5g Each for $331.49
Description & Specifications

Specifications

Formula Weight 166.18
Chemical Name or Material 3,4-Diaminobenzhydrazide
Assay 98%
Molecular Formula C7H10N4O
MDL Number MFCD00017069
CAS 103956-09-8
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 148° to 150°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.