missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,4-Diaminoanisole dihydrochloride, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 396990010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3, 4-Diaminoanisole dihydrochloride | |
97.5 to max. 102.5% min. (Argentometry) | |
C7H10N2O·2ClH | |
1g | |
Solubility in water: soluble. | |
COC1=CC(=C(C=C1)N)N.Cl.Cl | |
211.09 | |
211.09 |
59548-39-9 | |
Glass bottle | |
CH3OC6H3(NH2)2·2HCl | |
4-methoxybenzene-1,2-diamine dihydrochloride, 4-methoxy-o-phenylenediamine dihydrochloride, 3,4-diaminoanisole dihydrochloride, 1,2-benzenediamine, 4-methoxy-, dihydrochloride, 3,4-diaminoanisole 2hcl, 4-methoxy-phenylenediamine dihydrochloride, 4-methoxy-1,2-benzenediamine dihydrochloride, 4-methoxy-benzene-1,2-diamine dihydrochloride, 4-methoxy-1,2-phenylenediamine dihydrochloride, 4-methoxy-l,2-phenylene diamine dihydrochloride | |
SXCHMHOBHJOXGC-UHFFFAOYSA-N | |
4-methoxybenzene-1,2-diamine;dihydrochloride | |
3085161 | |
98% |
Chemical Identifiers
59548-39-9 | |
211.09 | |
4-methoxybenzene-1,2-diamine dihydrochloride, 4-methoxy-o-phenylenediamine dihydrochloride, 3,4-diaminoanisole dihydrochloride, 1,2-benzenediamine, 4-methoxy-, dihydrochloride, 3,4-diaminoanisole 2hcl, 4-methoxy-phenylenediamine dihydrochloride, 4-methoxy-1,2-benzenediamine dihydrochloride, 4-methoxy-benzene-1,2-diamine dihydrochloride, 4-methoxy-1,2-phenylenediamine dihydrochloride, 4-methoxy-l,2-phenylene diamine dihydrochloride | |
4-methoxybenzene-1,2-diamine;dihydrochloride |
C7H10N2O·2ClH | |
SXCHMHOBHJOXGC-UHFFFAOYSA-N | |
3085161 | |
COC1=CC(=C(C=C1)N)N.Cl.Cl |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 261-801-
RUO â Research Use Only