Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Diamino-5-bromopyridine, 96%

Chemical Identifiers

Chemical Name or Material 3,4-Diamino-5-bromopyridine
CAS 4635-08-9
Formula Weight 188.03
MDL Number MFCD01692479
UN Number UN2811
Synonym 5-Bromo-3,4-diaminopyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6682903

  SDS
alfa aesar™
H6682903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66829MD

  SDS
alfa aesar™
H66829MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 188.03
Melting Point 138° to 145°C
MDL Number MFCD01692479
CAS 4635-08-9
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 3,4-Diamino-5-bromopyridine
Physical Form Solid
Assay 96%
Synonym 5-Bromo-3,4-diaminopyridine
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Sensitivity Moisture & Light Sensitive
Color White to off-white to yellow
Quantity 1g
UN Number UN2811
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number US4000000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.