missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Diacetoxybenzonitrile, 97%

$96.07 - $382.45

Chemical Identifiers

CAS 203626-45-3
Molecular Formula C11H9NO4
Molecular Weight (g/mol) 219.196
InChI Key OYRIDYHXQXSNCH-UHFFFAOYSA-N
Synonym 3,4-diacetoxybenzonitrile, 3,4-diacetoxy benzonitrile, 4-cyano-1,2-phenylene diacetate, 2-acetyloxy-5-cyanophenyl acetate
PubChem CID 24721686
IUPAC Name (2-acetyloxy-4-cyanophenyl) acetate
SMILES CC(=O)OC1=C(C=C(C=C1)C#N)OC(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6620103 View Documents Alfa Aesar™
H6620103
1g Each for $96.07
 
AAH6620106 View Documents Alfa Aesar™
H6620106
5g Each for $382.45
 
Specifications

Chemical Identifiers

203626-45-3
219.196
3,4-diacetoxybenzonitrile, 3,4-diacetoxy benzonitrile, 4-cyano-1,2-phenylene diacetate, 2-acetyloxy-5-cyanophenyl acetate
(2-acetyloxy-4-cyanophenyl) acetate
C11H9NO4
OYRIDYHXQXSNCH-UHFFFAOYSA-N
24721686
CC(=O)OC1=C(C=C(C=C1)C#N)OC(=O)C

Specifications

White
1g
C11H9NO4
3,4-diacetoxybenzonitrile, 3,4-diacetoxy benzonitrile, 4-cyano-1,2-phenylene diacetate, 2-acetyloxy-5-cyanophenyl acetate
CC(=O)OC1=C(C=C(C=C1)C#N)OC(=O)C
219.196
219.19
97%
87°C
203626-45-3
UN3439
OYRIDYHXQXSNCH-UHFFFAOYSA-N
(2-acetyloxy-4-cyanophenyl) acetate
24721686
Crystalline Solid
3,4-Diacetoxybenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.