missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Diacetoxybenzamidoxime, 97%

Manufacturer:  Alfa Aesar™ H54541MD

 View more versions of this product

Catalog No. AAH54541MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3,4-Diacetoxybenzamidoxime
1858264-20-6
MFCD20265286
CC(=O)OC1=C(C=C(C=C1)C(=NO)N)OC(=O)C
252.226
252.23
250mg
C11H12N2O5
DUQSSXZZHVNOOD-UHFFFAOYSA-N
[2-acetyloxy-4-[(Z)-N'-hydroxycarbamimidoyl]phenyl] acetate
121235395
97%

Chemical Identifiers

1858264-20-6
252.226
DUQSSXZZHVNOOD-UHFFFAOYSA-N
[2-acetyloxy-4-[(Z)-N'-hydroxycarbamimidoyl]phenyl] acetate
C11H12N2O5
MFCD20265286
121235395
CC(=O)OC1=C(C=C(C=C1)C(=NO)N)OC(=O)C
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents