Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Diacetamidobenzoic acid, 97%

$43.73 - $133.00

Chemical Identifiers

Chemical Name or Material 3,4-Diacetamidobenzoic acid
CAS 205873-56-9
Molecular Formula C11H12N2O4
Formula Weight 236.23
MDL Number MFCD00238577
Synonym 3,4-Bis(acetamido)benzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2189006

  SDS
alfa aesar™
B2189006
5g Each for $133.00

AAB2189003

  SDS
alfa aesar™
B2189003
1g Each for $43.73
Description & Specifications

Specifications

Formula Weight 236.23
Chemical Name or Material 3,4-Diacetamidobenzoic acid
Assay 97%
Molecular Formula C11H12N2O4
MDL Number MFCD00238577
CAS 205873-56-9
Synonym 3,4-Bis(acetamido)benzoic acid
Melting Point 224° to 228°C
Quantity 5g