missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Di-O-acetyl-D-glucal, 97%

Manufacturer:  Alfa Aesar™ H60585MD

 View more versions of this product

Catalog No. AAH60585MD



Specifications

Specifications

3,4-Di-O-acetyl-D-glucal
82295-98-5
MFCD22989007
LBVCUTFITQDWSU-BBBLOLIVSA-N
[(2R,3S,4R)-3-acetyloxy-2-(hydroxymethyl)-3,4-dihydro-2H-pyran-4-yl] acetate
11064237
97%
250mg
C10H14O6
2r,3s,4r-4-acetyloxy-2-hydroxymethyl-3,4-dihydro-2h-pyran-3-yl acetate
CC(=O)OC1C=COC(C1OC(=O)C)CO
230.216
230.21

Chemical Identifiers

82295-98-5
230.216
LBVCUTFITQDWSU-BBBLOLIVSA-N
11064237
CC(=O)OC1C=COC(C1OC(=O)C)CO
C10H14O6
MFCD22989007
2r,3s,4r-4-acetyloxy-2-hydroxymethyl-3,4-dihydro-2h-pyran-3-yl acetate
[(2R,3S,4R)-3-acetyloxy-2-(hydroxymethyl)-3,4-dihydro-2H-pyran-4-yl] acetate
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C

Documents