missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ 3,4-Di-O-acetyl-6-O-tert-butyldimethylsilyl-D-glucal, 97%
Manufacturer: Alfa Aesar™ H60900MD
Specifications
3,4-Di-O-acetyl-6-O-tert-butyldimethylsilyl-D-glucal | |
−7° (c=1.4 in Chloroform) | |
117136-33-1 | |
MFCD01321277 | |
344.48 |
>113°C (235°F) | |
250mg | |
C16H28O6Si | |
2r,3s,4r-2-tert-butyldimethylsilyl oxy methyl-3,4-dihydro-2h-pyran-3,4-diyl diacetate | |
97% |
Chemical Identifiers
117136-33-1 | |
C16H28O6Si | |
MFCD01321277 | |
2r,3s,4r-2-tert-butyldimethylsilyl oxy methyl-3,4-dihydro-2h-pyran-3,4-diyl diacetate |
Safety and Handling
Recommended Storage : 2° to 8°C