missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Di-O-acetyl-6-O-tert-butyldimethylsilyl-D-glucal, 97%

$62.68 - $188.04

Chemical Identifiers

CAS 117136-33-1
Molecular Formula C16H28O6Si
MDL Number MFCD01321277
Synonym 2r,3s,4r-2-tert-butyldimethylsilyl oxy methyl-3,4-dihydro-2h-pyran-3,4-diyl diacetate
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60900MD View Documents Alfa Aesar™
H60900MD
250mg Each for $62.68
 
AAH6090003 View Documents Alfa Aesar™
H6090003
1g Each for $188.04
 
Specifications

Chemical Identifiers

117136-33-1
C16H28O6Si
MFCD01321277
2r,3s,4r-2-tert-butyldimethylsilyl oxy methyl-3,4-dihydro-2h-pyran-3,4-diyl diacetate

Specifications

1.044
−7° (c=1.4 in Chloroform)
250mg
C16H28O6Si
2r,3s,4r-2-tert-butyldimethylsilyl oxy methyl-3,4-dihydro-2h-pyran-3,4-diyl diacetate
97%
>113°C (235°F)
1.456
117136-33-1
MFCD01321277
344.48
3,4-Di-O-acetyl-6-O-tert-butyldimethylsilyl-D-glucal
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Keep cold