missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-[(4-Chlorophenylsulfonyl)methyl]benzamidoxime

$546.45 - $546.45

Chemical Identifiers

CAS 583059-52-3
Molecular Formula C14H13ClN2O3S
Formula Weight 324.79
MDL Number MFCD08060949
Synonym 3-([(4-Chlorophenyl)sulfonyl]methyl)-N-hydroxybenzenecarboximidamide
Chemical Name or Material 3-[(4-Chlorophenylsulfonyl)methyl]benzamidoxime
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5063306 SDS Alfa Aesar™
H5063306
5g Each for $546.45
AAH5063303 SDS Alfa Aesar™
H5063303
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

583059-52-3
324.79
3-([(4-Chlorophenyl)sulfonyl]methyl)-N-hydroxybenzenecarboximidamide
C14H13ClN2O3S
MFCD08060949
3-[(4-Chlorophenylsulfonyl)methyl]benzamidoxime

Specifications

5g
C14H13ClN2O3S
MFCD08060949
3-[(4-Chlorophenylsulfonyl)methyl]benzamidoxime
583059-52-3
324.79
3-([(4-Chlorophenyl)sulfonyl]methyl)-N-hydroxybenzenecarboximidamide

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning