Please login to your online account to display your discounted pricing

3-(4-Chlorophenyl)propan-1-ol, 95%, Maybridge

Chemical Identifiers

Chemical Name or Material 3-(4-Chlorophenyl)propan-1-ol
CAS 6282-88-8
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C9H11ClO
Linear Formula ClC6H4(CH2)3OH
Formula Weight 170.64
MDL Number MFCD00831102
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

KM10071EA

  SDS
fisher scientific company
KM10071EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 170.64
Quantity 10g
Assay 95%
CAS Min % 95.0
Chemical Name or Material 3-(4-Chlorophenyl)propan-1-ol
Packaging Amber Glass Bottle
Assay Percent Range 95%
Molecular Formula C9H11ClO
Linear Formula ClC6H4(CH2)3OH
MDL Number MFCD00831102
CAS 6282-88-8