Please login to your online account to display your discounted pricing

3-(4-Chlorophenyl)propan-1-ol, 95%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  KM10071EA

 View more versions of this product

Catalog No. KM10071EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 170.64
Quantity 10g
Assay 95%
CAS Min % 95.0
Chemical Name or Material 3-(4-Chlorophenyl)propan-1-ol
Packaging Amber Glass Bottle
Assay Percent Range 95%
Molecular Formula C9H11ClO
Linear Formula ClC6H4(CH2)3OH
MDL Number MFCD00831102
CAS 6282-88-8

Specifications

Chemical Name or Material 3-(4-Chlorophenyl)propan-1-ol
CAS 6282-88-8
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C9H11ClO
Linear Formula ClC6H4(CH2)3OH
Formula Weight 170.64
MDL Number MFCD00831102