Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Chlorophenyl)glutaric acid, 98%


Quantity 5g

Manufacturer: alfa aesar™  L1869306

 View more versions of this product

Catalog No. AAL1869306

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 242.66
Chemical Name or Material 3-(4-Chlorophenyl)glutaric acid
CAS 35271-74-0
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Molecular Formula C11H11ClO4
MDL Number MFCD00190249
Beilstein 1976828
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 164° to 168°C
Quantity 5g

Specifications

Chemical Name or Material 3-(4-Chlorophenyl)glutaric acid
CAS 35271-74-0
Molecular Formula C11H11ClO4
Formula Weight 242.66
MDL Number MFCD00190249
Beilstein 1976828

Applications
It is employed in an efficient synthesis of (R)- and (S)-baclofen via desymmetrization.

Solubility
Soluble in dimethyl sulfoxide. Slightly soluble in methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Accessories

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.