Please login to your online account to display your discounted pricing

3-(4-chlorophenyl)-4-cyano-5-(methylthio)thiophene-2-carboxylic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 3-(4-Chlorophenyl)-4-cyano-5-(methylthio)thiophene-2-carboxylic acid
CAS 116525-66-7
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C13H8ClNO2S2
Formula Weight 309.8
MDL Number MFCD00112478
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

KM01185DA

  SDS
fisher scientific company
KM01185DA
Amber glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

KM01185CB

  SDS
fisher scientific company
KM01185CB
Amber glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 309.8
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 3-(4-Chlorophenyl)-4-cyano-5-(methylthio)thiophene-2-carboxylic acid
Packaging Amber glass bottle
Assay Percent Range 97%
Molecular Formula C13H8ClNO2S2
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful in contact with skin.
MDL Number MFCD00112478
CAS 116525-66-7
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.