Please login to your online account to display your discounted pricing

3-(4-Chlorophenyl)-1-methyl-1H-pyrazol-5-amine, ≥97%, Maybridge


Quantity 10g

Manufacturer: fisher scientific company  KM04038EA

 View more versions of this product

Catalog No. KM04038EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 207.66
Quantity 10g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 3-(4-Chlorophenyl)-1-methyl-1H-pyrazol-5-amine
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Assay Percent Range 97%
Molecular Formula C10H10ClN3
MDL Number MFCD00084872
CAS 126417-82-1
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3-(4-Chlorophenyl)-1-methyl-1H-pyrazol-5-amine
CAS 126417-82-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H10ClN3
Formula Weight 207.66
MDL Number MFCD00084872
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.