Please login to your online account to display your discounted pricing

3-(4-Chlorophenoxy)pentane-2,4-dione, Tech., Maybridge

Chemical Identifiers

Chemical Name or Material 3-(4-Chlorophenoxy)pentane-2, 4-dione
Assay Tech.
Molecular Formula C11H11ClO3
Linear Formula ClC6H4OCHC(O)CH32
Formula Weight 226.66
MDL Number MFCD00204239
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

SPB03987DA

  SDS
fisher scientific company
SPB03987DA
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SPB03987EA

  SDS
fisher scientific company
SPB03987EA
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 226.66
Purity Grade Technical Grade
Quantity 1g
Assay Tech.
MDL Number MFCD00204239
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 3-(4-Chlorophenoxy)pentane-2, 4-dione
CAS 31168-10-2
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Linear Formula ClC6H4OCHC(O)CH32
Molecular Formula C11H11ClO3
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.