missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-[4-(Chloromethyl)phenoxy]-N,N-dimethylpropylamine hydrochloride, 90%, Maybridge

Chemical Identifiers

CAS 769123-68-4
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Molecular Formula C12H18ClNO·ClH
Formula Weight 264.19
MDL Number MFCD09702396
Chemical Name or Material 3-[4-(Chloromethyl)phenoxy]-N, N-dimethylpropylamine hydrochloride
Assay 90%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC59574DA SDS Fisher Scientific Company
CC59574DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

769123-68-4
100.0
C12H18ClNO·ClH
MFCD09702396
90%
89.5
90%
264.19
3-[4-(Chloromethyl)phenoxy]-N, N-dimethylpropylamine hydrochloride

Specifications

769123-68-4
100.0
90%
C12H18ClNO·ClH
MFCD09702396
90%
89.5
Amber Glass Bottle
1g
264.19
3-[4-(Chloromethyl)phenoxy]-N, N-dimethylpropylamine hydrochloride

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning