Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-[(4-Chlorobenzyl)thio]-1H-1,2,4-triazole

$99.89 - $99.89

Chemical Identifiers

Chemical Name or Material 3-[(4-Chlorobenzyl)thio]-1H-1,2,4-triazole
CAS 134796-34-2
Molecular Formula C9H8ClN3S
Formula Weight 225.7
MDL Number MFCD01211119
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH50647MD

  SDS
alfa aesar™
H50647MD
250mg Each for $99.89
Description & Specifications

Specifications

Formula Weight 225.7
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 3-[(4-Chlorobenzyl)thio]-1H-1,2,4-triazole
Molecular Formula C9H8ClN3S
MDL Number MFCD01211119
CAS 134796-34-2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 250mg
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.