Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Chlorobenzoyl)propionic acid, 98%

$213.96 - $213.96

Chemical Identifiers

Chemical Name or Material 3-(4-Chlorobenzoyl)propionic acid
CAS 3984-34-7
Molecular Formula C10H9ClO3
Formula Weight 212.63
MDL Number MFCD00002794
Beilstein 2104409
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1899622

  SDS
alfa aesar™
A1899622
100g Each for $213.96
Description & Specifications

Specifications

Formula Weight 212.63
Quantity 100g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 3-(4-Chlorobenzoyl)propionic acid
Beilstein 2104409
Health Hazard 1 Warning
Assay 98%
Molecular Formula C10H9ClO3
Melting Point 128° to 130°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00002794
CAS 3984-34-7
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.