Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Chlorobenzoyl)acrylic acid, 99%

$41.88 - $170.86

Chemical Identifiers

Chemical Name or Material 3-(4-Chlorobenzoyl)acrylic acid
CAS 6269-33-6
Molecular Formula C10H7ClO3
Formula Weight 210.62
MDL Number MFCD00041465
Beilstein 2719881
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1146706

  SDS
alfa aesar™
L1146706
5g Each for $41.88

AAL1146714

  SDS
alfa aesar™
L1146714
25g Each for $170.86
Description & Specifications

Specifications

Formula Weight 210.62
Chemical Name or Material 3-(4-Chlorobenzoyl)acrylic acid
Assay 99%
MDL Number MFCD00041465
CAS 6269-33-6
Beilstein 2719881
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 157° to 159°C
Molecular Formula C10H7ClO3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.