missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Chlorobenzoyl)acrylic acid, 99%

Manufacturer:  Alfa Aesar™ L1146706

 View more versions of this product

Catalog No. AAL1146706



Specifications

Specifications

3-(4-Chlorobenzoyl)acrylic acid
5g
C10H7ClO3
2719881
VQVQEUFKSRHRCT-AATRIKPKSA-N
(E)-4-(4-chlorophenyl)-4-oxobut-2-enoic acid
671716
99%
157°C to 159°C
6269-33-6
MFCD00041465
3-4-chlorobenzoyl acrylic acid, p-chlorobenzoylacrylic acid, trans-3-4-chlorobenzoyl acrylic acid, e-4-4-chlorophenyl-4-oxo-2-butenoic acid, 2e-4-4-chlorophenyl-4-oxobut-2-enoic acid, acide e 4-4-chlorophenyl 4-oxo 2-butenoique french, 2-butenoic acid, 4-4-chlorophenyl-4-oxo-, e, 4-4-chlorophenyl-4-oxo-2-butenoic acid, e-4-4-chlorophenyl-4-oxobut-2-enoic acid, e-4-4-chlorophenyl-4-oxo-but-2-enoic acid
C1=CC(=CC=C1C(=O)C=CC(=O)O)Cl
210.613
210.62

Chemical Identifiers

6269-33-6
210.613
VQVQEUFKSRHRCT-AATRIKPKSA-N
671716
C1=CC(=CC=C1C(=O)C=CC(=O)O)Cl
C10H7ClO3
MFCD00041465
3-4-chlorobenzoyl acrylic acid, p-chlorobenzoylacrylic acid, trans-3-4-chlorobenzoyl acrylic acid, e-4-4-chlorophenyl-4-oxo-2-butenoic acid, 2e-4-4-chlorophenyl-4-oxobut-2-enoic acid, acide e 4-4-chlorophenyl 4-oxo 2-butenoique french, 2-butenoic acid, 4-4-chlorophenyl-4-oxo-, e, 4-4-chlorophenyl-4-oxo-2-butenoic acid, e-4-4-chlorophenyl-4-oxobut-2-enoic acid, e-4-4-chlorophenyl-4-oxo-but-2-enoic acid
(E)-4-(4-chlorophenyl)-4-oxobut-2-enoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents