Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-[4-Chloro-3-(trifluoromethyl)phenyl]propionic acid, 97%

$212.46 - $212.46

Chemical Identifiers

Chemical Name or Material 3-[4-Chloro-3-(trifluoromethyl)phenyl]propionic acid
CAS 900027-15-8
Molecular Formula C10H8ClF3O2
Formula Weight 252.62
MDL Number MFCD06823954
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3246003

  SDS
alfa aesar™
H3246003
1g Each for $212.46
Description & Specifications

Specifications

Formula Weight 252.62
Assay 97%
Chemical Name or Material 3-[4-Chloro-3-(trifluoromethyl)phenyl]propionic acid
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 77° to 82°C
CAS 900027-15-8
Quantity 1g
MDL Number MFCD06823954
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Molecular Formula C10H8ClF3O2
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.